Latent Fingerprinting Supplies
Catalog #: 470020-198
Supplier: FORENSICS SOURCE
Specifications
- Description:The Breeze™ Fingerprint Brush
- Cat. no.:470020-198
- Length:6 1/4"
- Supplier No.:1-0300
Specifications
About this item
Develop and study otherwise scarcely visible fingerprints using high-quality fingerprint brushes.
- Different fingerprinting brushes to suit your preferences
- Supplies Demonstrate the minute differences in individual fingerprints
- Camel Brush Ideal for General Conditions
- Zephyr Brush Retains a Large Amount of Powder
- Brush Length: 61/4"
For general conditions, the camel hair brush is ideal.
The Zephyr fiberglass brush retains a larger amount of powder and applies it more evenly to provide clear prints in any circumstance.
Customers Who Bought This Also Bought