Order Entry
ContactUsLinkComponent
BRUSH T-TUBE SEMI-MICRO 5-7/8 WLS10007
Nylon Semimicro Test Tube Brush
Catalog #: 470016-166
Supplier:  Mill-Rose
BRUSH T-TUBE SEMI-MICRO 5-7/8 WLS10007
Nylon Semimicro Test Tube Brush
Catalog #: 470016-166
Supplier:  Mill-Rose

Specifications

  • Bristle Color:
    Black
  • Bristle Material:
    Nylon
  • Description:
    Semimicro Test Tube Brush, Each
  • Cat. no.:
    470016-166
  • Supplier No.:
    73200B

Specifications

About this item

For test tubes 5 to 11 mm in diameter. Total length: 14.6 cm; length of fiber section: 7 cm; diameter: 1.3 cm.

Customers Who Bought This Also Bought