Order Entry
ContactUsLinkComponent
SCALPEL HANDLE SS 4 140MM
Lock-On Scalpel Handles
Catalog #: 470007-550
SCALPEL HANDLE SS 4 140MM
Lock-On Scalpel Handles
Catalog #: 470007-550

Specifications

  • Length:
    140 mm (5.5")
  • Description:
    Lock-On Scalpel Handle No. 4
  • Material:
    Stainless Steel
  • Cat. no.:
    470007-550
  • Supplier No.:
    140707

Specifications

About this item

The traditional surgical handle.

  • Lock on with secure hold
  • Safely and easily change blades
  • Wide variety of blade types

These stainless steel handles with lock-on device make changing blades easy.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought