Order Entry
ContactUsLinkComponent
MIXER LABORATORY BLENDER
Laboratory Blender/Mixer
Catalog #: 470006-518
Supplier:  CONAIR CORPORATION
MIXER LABORATORY BLENDER
Laboratory Blender/Mixer
Catalog #: 470006-518
Supplier:  CONAIR CORPORATION

Specifications

  • Cat. no.:
    470006-518
  • Electrical:
    155 VAC
  • Diameter ×L:
    17.9 dia. x 39.1 L cm (7 x 14.5" H)
  • Supplier No.:
    700G

Specifications

About this item

Heavy-duty.

  • Designed for shredding, blending, and emulsification
  • Features stainless steel blades and lubricated bearings
Customers Who Bought This Also Bought