Bibulous Paper
Catalog #: 470005-600
Supplier: Raylabcon
Specifications
- Dimensions:4 x 6"
- Description:Ward's® Bibulous Paper
- Cat. no.:470005-600
- Supplier No.:20105000
Specifications
About this item
Common blotting and cleaning paper.
- Quality slide making supplies
- Helpful for maintaining cleanliness of slides
Use this bibulous paper to clean glass surfaces of dust and lint without leaving streaks. It also blots excess moisture from slides containing living materials. Staple-bound booklet of 50 sheets.
Customers Who Bought This Also Bought