Order Entry
ContactUsLinkComponent
POLYHEDRAL ALCOHOL BURNER 125ML
Polyhedral Alcohol Burner
Catalog #: 470020-698
Small Business Enterprise
POLYHEDRAL ALCOHOL BURNER 125ML
Polyhedral Alcohol Burner
Catalog #: 470020-698
Small Business Enterprise

Specifications

  • Description:
    Polyhedralburner
  • Cat. no.:
    470020-698
  • Supplier No.:
    2066-11

Specifications

About this item

Position the Flame at Just the Right Angle.

  • Capacity: 125 mL
  • Height: 4"

The polyhedral shape of this heavy-duty glass burner allows you to position the flame at just the right angle for different tasks. It comes with a nickel-plated snuffer cap and one wick. Uses 1/4" (6mm) thick replacement wicks.

Customers Who Bought This Also Bought