Ward's® Syringe-style Micropipettes
Catalog #: 470019-918
Supplier: IPM SCIENTIFIC, INC.
Specifications
- Description:Micropet Rack (holds 6)
- Cat. no.:470019-918
- Supplier No.:11003-010W
Specifications
About this item
Economical choice for basic applications.
- Measures 10, 20, 50, 100, and 200 μl
- Comes with two tips and a tip gauge
- Optional rack/stand keeps equipment safe between use
Measure and transfer small amounts of liquid with our inexpensive micropipette. Each tip is graduated for accurate measurements, and the built-in stop at 100 μl prevents any overflow into the tip adapter.
Customers Who Bought This Also Bought