ContactUsLinkComponent
DENSITOMETER
Densitometer
Catalog #: 470018-578
DENSITOMETER
Densitometer
Catalog #: 470018-578
Specifications
  • Description:
    Densitometer
  • Cat. no.:
    470018-578
  • Supplier No.:
    101255
Specifications
About this item
Estimate vegetation cover like the professionals.
  • Aligned with GLOBE protocols
The combined horizontal and vertical vegetation sampling available from this valuable tool enables your students to collect resource information both across the landscape and at different canopy levels. Using a mirror to project a view of the sample location point in the canopy above to the person holding the densitometer, it will provide accurate, objective, and repeatable measurements of cover for any element in a forested environment. Simply align the two bubble-line levels mounted inside to get an exact vertical line-of-sight into the canopy, then record the data point viewed through the centered cross hairs. Size: 3 1/2"L x 1 5/8"W x 5"H.
Customers Who Bought This Also Bought