Order Entry
ContactUsLinkComponent
LAMOTTE DISSOLVED OXYGEN TRACER
LaMotte® Dissolved Oxygen TRACER
Catalog #: 470012-536
Supplier:  LaMotte
LAMOTTE DISSOLVED OXYGEN TRACER
LaMotte® Dissolved Oxygen TRACER
Catalog #: 470012-536
Supplier:  LaMotte
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    LaMotte® Dissolved Oxygen TRACER
  • Cat. no.:
    470012-536
  • Supplier No.:
    1761

Specifications

About this item

Measure oxygen concentration and percent saturation

  • A great way to monitor a variety of common water quality factors without having to measure reagents
  • Compare your readings to a handy color comparator chart printed right on the storage vial
  • The six-way strips test: chlorine 0 to 10 ppm, pH 4 to 10, hardness 0 to 400 ppm, nitrate 0 to 50 ppm, and nitrite 0 to 10 ppm
  • The five-way strips test: nitrate 0 to 200 ppm, nitrite 0 to 10 ppm, pH 6 to 9, alkalinity 0 to 240 ppm, and hardness 0 to 180 ppm

This accurate instrument features a large memory to hold up to 25 data sets with both dissolved oxygen (DO) and temperature readings. Oxygen level is displayed as percent saturation from 0 to 200% or concentration from 0 to 20.00 ppm (mg/l). Adjustable altitude compensation is 0 to 20,000' in 1,000' increments, and adjustable salinity compensation is from 0 to 50 ppt. It self-calibrates when turned on, includes auto-power off and a low battery indicator. Includes DO electrode, protective sensor cap, spare membrane cap, electrolyte, four 1.5 V SR44 W batteries, and a 48 neckstrap.

DO Range: 0 to 200%; 0 to 20.00 ppm. Resolution: 0.1%. Accuracy: ±2%; ±0.4ppm.
Temperature Range: 0 to 50 °C; 32 to 122 °F. Resolution: 0.1 °C/°F. Accuracy: ±1 °C/1.8 °F.

Customers Who Bought This Also Bought