Chemistry Gram Stain Kit
Catalog #: 470010-662
Supplier: Ward's Science
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Cat. no.:470010-662
- Supplier No.:GG0221-KIT
- No. of Uses:50 slides
Specifications
About this item
The Chemistry Gram Stain Kit makes gram straining easy and fast.
- Shelf Life of 36 Months
- Students Compare Microorganisms and Their Characteristics
- No Chemical Mixing
- Multiple Solutions Included
- Fast and Simple Process
It lets you compare microorganisms and their characteristics and doesn't require chemical mixing.
Certifications: Note:This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Ordering information: Kit No. 470010-662 contains
30ml crystal violet
30ml iodine potassium iodide
30ml safranin O
100ml 95% ethanol
Kit No. 470302-350 contains
500ml crystal violet
500ml iodine potassium iodide
500ml safranin O
2 X 1000ml 95% ethanol
30ml crystal violet
30ml iodine potassium iodide
30ml safranin O
100ml 95% ethanol
Kit No. 470302-350 contains
500ml crystal violet
500ml iodine potassium iodide
500ml safranin O
2 X 1000ml 95% ethanol
Customers Who Bought This Also Bought