Order Entry
ContactUsLinkComponent
P/B SPIROMETER DISPOSABLE MOUTHPIECES
Student Wet Spirometer, 9 L
Catalog #: 470007-992
Supplier:  PHIPPS & BIRD INC.
P/B SPIROMETER DISPOSABLE MOUTHPIECES
Student Wet Spirometer, 9 L
Catalog #: 470007-992
Supplier:  PHIPPS & BIRD INC.

Specifications

  • Description:
    Disposable Mouthpieces
  • Cat. no.:
    470007-992
  • Outer Diameter:
    2.2 cm
  • Supplier No.:
    7085-301

Specifications

About this item

Increased Volume Size to Accommodate Students with Larger Lung Capacities

Measure Tidal Volume (TV), Inspiratory Capacity (IC), Inspiratory Reserve Volume (IRV), Expiratory Reserve Volume (ERV), and Vital Capacity (VC) with this spirometer that features large, easy-to-read scale graduations. Constructed of durable polyethylene and aluminum, it comes with 100 disposable mouthpieces, a one way plastic valve for sanitary classroom use, instructions, and a lab manual of experiments. Size: 30"H x 18"L x 18"W.

Customers Who Bought This Also Bought