Order Entry
ContactUsLinkComponent
ECONOMY ALCOHOL BURNER 120ML ROUND
Economy Alcohol Burners
Catalog #: 470002-066
Supplier:  C & A Scientific
ECONOMY ALCOHOL BURNER 120ML ROUND
Economy Alcohol Burners
Catalog #: 470002-066
Supplier:  C & A Scientific

Specifications

  • Volume:
    120 ml (4 oz.)
  • Description:
    Economy alcohol burner
  • Cat. no.:
    470002-066
  • Supplier No.:
    97-5313

Specifications

About this item

Improved Design.

  • Round body with secure screw-on top
  • Brass snuffer cap for extinguishing flame and preventing odor and evaporation

Alcohol not included.

Customers Who Bought This Also Bought