WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Catalog #: 30618-468
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Septum:Butyl stopper
- Description:Septa, bulk
- Cat. no.:30618-468
- Supplier No.:20-0025-B
Specifications
About this item
These septa are used for 20 mm headspace vial crimp seals.
Designed for optimum performance in a variety of applications and heat ranges.
Customers Who Bought This Also Bought