Order Entry
ContactUsLinkComponent
Eps Insulated Shipper Foam/Corrugate Id 15-13/16In X 12-13/16In X 8-1/2In W/ 1In Wall
Insulated Shippers, Expanded Polystyrene (EPS), Foam Only and with Corrugate, Sonoco ThermoSafe
Catalog #: 89413-404
Supplier:  Sonoco Thermosafe
Eps Insulated Shipper Foam/Corrugate Id 15-13/16In X 12-13/16In X 8-1/2In W/ 1In Wall
Insulated Shippers, Expanded Polystyrene (EPS), Foam Only and with Corrugate, Sonoco ThermoSafe
Catalog #: 89413-404
Supplier:  Sonoco Thermosafe

Specifications

  • Description:
    Multi-purpose foam container assembled in corrugated carton
  • Thickness:
    2.5 cm (1")
  • Cat. no.:
    89413-404
  • Interior Dimensions:
    1513/16L×1213/16W×81/2H"
  • Exterior Dimensions:
    181/8L×151/8W×107/8H"
  • Supplier No.:
    658

Specifications

About this item

ThermoSafe's extensive line of EPS foam-only and EPS foam with corrugate insulated shippers offers the ideal solution for the safe storage and delivery of most refrigerated or frozen shipping products.

  • Large selection available
  • Excellent dry ice sublimation

A large selection of cost-effective, reusable EPS foam-only and EPS foam in corrugate stock containers are available. Excellent dry ice sublimation; average dry ice usage in 1-1/2" thick wall units is 5 lbs. over 24 hours and 3-4 lbs. thereafter. Standard 1-1/2" wall shippers are ideal for shipments of 24 to 48 hours. Thin wall shippers have 3/4 to 1-1/8" of EPS foam insulation. Ideal for shipments of 24 hours or less. Thick wall shippers have 1-3/4 to 2-1/4" of EPS insulation specifically designed for shipments of 48 hours or more.

Packaging: EPS foam with corrugate shippers are EPS foam containers in 200 lb. test corrugated carton.
Customers Who Bought This Also Bought