ContactUsLinkComponent
PIG BENCH PADS FLAT SHEET 100/CASE
Bench Pads, PIG®, New Pig
Catalog #: 89412-166
Supplier:  New Pig
PIG BENCH PADS FLAT SHEET 100/CASE
Bench Pads, PIG®, New Pig
Catalog #: 89412-166
Supplier:  New Pig
Specifications
  • W×L:
    12x15"
  • Description:
    PIG® Bench Pads, Flat Sheet
  • Cat. no.:
    89412-166
  • Supplier No.:
    WIP1600
Specifications
About this item
This PIG-exclusive absorbent is just right for benchtop jobs and small spill cleanup.
  • Fresh, disposable work surface for each new job
  • Lays flat and stays flat
  • No wrinkles or creases
Thicker than a wiper, thinner than a mat, this PIG-exclusive absorbent is just right for benchtop jobs and small spill cleanup. Affordable pads give you a fresh, disposable work surface for each new job without creating extra waste. Bench pad lays flat and stays flat; no wrinkles or creases which cause small parts to roll off your bench.
Customers Who Bought This Also Bought