Specifications
- Electrical:120 V
- Overall Dimensions:25.4D x 24.9W x 5.7H cm (10 x 93/4 x 21/4")
- Top Plate Dimensions:17.8D×20.3W cm (7×8")
- Supplier No.:F37015-0000
Specifications
About this item
Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1 °C) outside of the incubator.
The temperature remains uniform across the entire anodized aluminum warming plate, which measures 17.8Dx20.3W cm (7x8"). The unit's small footprint allows it to easily fit in a cell culture hood or on the workbench. The UV resistant polypropylene housing will not break down under germicidal UV lights.
- Reduces cell stress and allows enzymes like trypsin to work efficiently
- Can be used to maintain the warmth of reagents inside the hood
- Fits easily in cell culture hoods or on the workbench
- Temperature remains uniform across the entire anodized aluminum warming plate
- 15-minute count down timer keeps track of time sensitive applications
- UV resistant polypropylene housing will not break down under germicidal UV lights
- Water resistant, safe for cleaning with ethanol or IPA
Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1 °C) outside of the incubator.
Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures.
Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures.