Total Hardness Test Kit, Model HA-71A, Hach
Catalog #: CA11024-936
Supplier: Hach
CAS Number:
Specifications
- Chemical method:EDTA
- Description:Total Hardness Test Kit, Model HA-71A
- Model:HA-71A
- Method:Drop count titration/EDTA-powder pillows
- Case style:D
- Increments:smallest steps: 1 mg/L & 1 gpg
- Cat. no.:CA11024-936
- Range:1 - 20 gpg CaCO₃, 1 - 20 mg/l CaCO₃
- Supplier No.:145201
- No. of Tests:100
Specifications
About this item
Water hardness is caused almost entirely by calcium and magnesium ion.
-Utilizes drop count titration - a simple, economical titration method.
-Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
- Utilizes drop count titration a simple, economical titration method.
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Other di- and trivalent metals have a similar effect, but usually are not present in high enough concentration in potable waters to cause problems. Hardness increases soap consumption in laundries and causes scale in boilers. With the Model HA-71A test kit, simply count the number of drops required to change the solution color to determine hardness concentration. Utilizing drop count titration, this kit provides dual ranges and contains approximately 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Customers Who Bought This Also Bought