ContactUsLinkComponent
Pipette Tip 1-200Ul Yel Rk Ns
MBP 1 to 200 µl Pipette Tips, Thermo Scientific
Catalog #: 89404-796
Supplier:  Thermo Scientific
Pipette Tip 1-200Ul Yel Rk Ns
MBP 1 to 200 µl Pipette Tips, Thermo Scientific
Catalog #: 89404-796
Supplier:  Thermo Scientific
Specifications
  • Volume:
    1 - 200 µl
  • Tip Color:
    Yellow
  • Packed:
    96 Tip Rack with Lift-off Lid
  • Supplier No.:
    110-96R
Specifications
About this item
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
  • Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals
  • A mold release agent is NOT used in the manufacturing of these pipette tips
Autoclavable Tips & Racks: pipette tips and racks are autoclavable at a recommended temperature of 120°C at 15 pounds of pressure for 15 minutes. For use with but not limited to: Pipetman®, Costar® Onepette, Eppendorf®, Finnpipette®, Labpette™, Nichiryo, Oxford-Benchmate®, Rainin EDP2™, Socorex®, Titertek® Digital, Volac, and GeneMate®.
Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request.
Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.
A trace metal certificate is available upon request.
Customers Who Bought This Also Bought