ContactUsLinkComponent
ASCORBIC QUANT KIT 100TST/EA
Ascorbic Acid Test Method: Colorimetric With Test Strips 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L MQuant™, MilliporeSigma
Catalog #: CA1.10023.0001
Supplier:  MilliporeSigma
CAS Number:  
Ascorbic Acid Test Method: Colorimetric With Test Strips 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L MQuant™, MilliporeSigma
Catalog #: CA1.10023.0001
Supplier:  MilliporeSigma
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Chemical method:
    Phosphoromolybdenum blue
  • Description:
    Ascorbic Acid Test
  • Cat. no.:
    CA1.10023.0001
  • Range:
    50-100-200-300-500-700-1000-2000 mg/L
  • Supplier No.:
    1.10023.0001
  • No. of Tests:
    100 strips
Specifications
About this item
The ascorbic acid test is suitable for quickly measuring levels of natural ascorbic acid (vitamin C) in foods such as fruit and vegetable juices, soft drinks, and also to check how much ascorbic acid has been added to certain foods as a preservative or antioxidant.
MilliporeSigma # 1.10023.0001 replaces and is identical to MilliporeSigma # 10023-1
Customers Who Bought This Also Bought