Specifications
- Resolution:1.0
- Description:pH-Indicator strips, universal indicator, Non-bleeding, all-universal range
- Cat. no.:CA1.09535.0007
- pH Range:0 - 14
- pH Graduation:0-1.0-2.0-3.0-4.0-5.0-6.0-7.0-
8.0-9.0-10.0-11.0-12.0-13.0-14.0 - Supplier No.:1.09535.0007
- No. of Strips:600
Specifications
About this item
These strips provide a convenient method for accurate colorimetric pH indication.
The strips feature covalently bound indicator dyes so that they will not bleed even in strong alkaline solutions. Strips can be immersed in samples for extended periods, so that even weakly buffered solutions can be accurately tested without contaminating the sample. The universal strip tests the full 0–14 pH range with a sensitivity of 1 pH unit. Intermediate range indicators provide a sensitivity of 0.3–0.5 pH units and narrow range strips provide a sensitivity of 0.2–0.3 pH units.
MilliporeSigma # 1.09531.0001 replaces and is identical to MilliporeSigma # 9586-1.
MilliporeSigma # 1.09531.0007 replaces and is identical to MilliporeSigma # 9586-3.
MilliporeSigma # 1.09532.0001 replaces and is identical to MilliporeSigma # 9587-1.
MilliporeSigma # 1.09533.0001 replaces and is identical to MilliporeSigma # 9588-1.
MilliporeSigma # 1.09533.0007 replaces and is identical to MilliporeSigma # 9588-3.
MilliporeSigma # 1.09535.0001 replaces and is identical to MilliporeSigma # 9590-1.
MilliporeSigma # 1.09535.0007 replaces and is identical to MilliporeSigma # 9590-3.
MilliporeSigma # 1.09540.0001 replaces and is identical to MilliporeSigma # 9580-1.
MilliporeSigma # 1.09541.0001 replaces and is identical to MilliporeSigma # 9581-1.
MilliporeSigma # 1.09542.0001 replaces and is identical to MilliporeSigma # 9582-1.
MilliporeSigma # 1.09542.0007 replaces and is identical to MilliporeSigma # 9582-3.
MilliporeSigma # 1.09543.0001 replaces and is identical to MilliporeSigma # 9583-1.
MilliporeSigma # 1.09543.0007 replaces and is identical to MilliporeSigma # 9583-3.
MilliporeSigma # 1.09545.0001 replaces and is identical to MilliporeSigma # 9585-1.
MilliporeSigma # 1.09584.0001 replaces and is identical to MilliporeSigma # 9578-1.