Alcohol, anhydrous 94.0-96.0% (v/v), PHOTREX® for spectrophotometry, for liquid chromatography (denatured with 5% isopropanol and 5% methanol), J.T.Baker®
Catalog #: CAJT9229-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 64-17-5
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAJT9229-3
- Supplier No.:9229-03
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v):94.0 - 96.0 %
- Isopropyl Alcohol (2-propanol)(v/v):4.0 - 6.0 %
- Ethanol (v/v):88.5 - 92.5 %
- Methanol (v/v):3.5 - 5.5 %
- Appearance:Passes Test
- Color (APHA):≤5
- Residue after Evaporation:≤5 ppm
- Specific Gravity at 25°C:0.785 - 0.795
- Water (H₂O):≤0.1 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 210 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 220 nm:≤0.50
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 230 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 250 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 270 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400 nm:≤0.01
- Substances Reducing Permanganate:Passes Test
- Titrable Acid (µeq/g):
- Titrable Base (µeq/g):
- SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.:
- J.T.Baker®, Alcohol, anhydrous 94.0-96.0% (v/v), PHOTREX® for spectrophotometry, for liquid chromatography (denatured with 5% isopropanol and 5% methanol):
Specification Test Results
About this item
Synonyms: Reagent alcohol
:Alcohol
Boiling Pt:78.3 °C (1013 hPa)
Melting Pt:–114 °C
Density:0.7895 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:94,0-96,0
:Management of Change (MOC) category = N
Flash Pt:13 °C
:Boiling Pt:
:Boiling Pt: 78.3 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –114 °C
:Flash Pt:
:Flash Pt: 13 °C
EINECS:200-578-6
:78.3 °C (1013 hPa)
Storage Temperature:Ambient
UN:1987
REACH:01-2119457610-43
:3
:II
:13 °C
:GHS02
:GHS07
:GHS08
:M.O.C. Category N
:12,03806
Customers Who Bought This Also Bought