Specifications
- Description:Needle
- Cat. no.:CABD368607L
- Needle Length:3.175 cm (11/4")
- Needle Gauge:21 G
- Supplier No.:368607
Specifications
About this item
These sterile, safety-engineered, multi-sample blood collection needles offer a simple, effective way to collect blood while reducing the possibility of needlestick injuries
These sterile, safety-engineered, multi-sample blood collection needles offer a simple, effective way to collect blood while reducing the possibility of needlestick injuries
They feature a patented safety shield that allows for one-handed activation to cover the needle completely upon withdrawal from the vein, and confirm proper activation with an audible click. The safety shield is already aligned with the bevel of the needle, so no extra assembly or adjustment is required by the user. The intuitive design of the advanced protection technology means there is no need to change the standard, one-handed venipuncture technique when using these needles. Needles with pre-attached holders help facilities remain single-use holder compliant.