Order Entry
ContactUsLinkComponent
LANCET QUICKHEEL INFANT PK50
BD Microtainer® Quikheel™ Lancets, BD Biosciences
Catalog #: CABD368101L
Supplier:  Discovery Labware
LANCET QUICKHEEL INFANT PK50
BD Microtainer® Quikheel™ Lancets, BD Biosciences
Catalog #: CABD368101L
Supplier:  Discovery Labware
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Color:
    Teal
  • Description:
    Infant Lancet
  • Cat. no.:
    CABD368101L
  • Incision Depth:
    1.0 mm
  • Incision Length:
    2.5 mm
  • Supplier No.:
    368101

Specifications

About this item

Lancets are gentle and safety-engineered blood sampling devices designed for heel-sticks in premature infants, newborns, and babies

Lancets are gentle and safety-engineered blood sampling devices designed for heel-sticks in premature infants, newborns, and babies

An extra-thin surgical steel blade provides a fine, clean surgical incision. An automatic, permanently retractable blade minimizes possible injury or reuse. A tapered contact surface allows a clear view of the site location while a finger-contoured design provides a steady and stable grip. The instant trigger action eliminates the need for safety tabs to prevent blade release. Lancets are color-coded for easy selection. Sterile.

Customers Who Bought This Also Bought