Order Entry
ContactUsLinkComponent
Spec Clor Ozone 0-0.75Mg/L
SpecCheck Gel Secondary Standard Set, Ozone, Mid-Range, Hach
Catalog #: CA97026-524
Supplier:  Hach
CAS Number:  
Spec Clor Ozone 0-0.75Mg/L
SpecCheck Gel Secondary Standard Set, Ozone, Mid-Range, Hach
Catalog #: CA97026-524
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    0 - 0.75 mg/L O₃
  • Description:
    SpecCheck Gel Secondary Standard Set, Ozone, Mid-Range
  • Method:
    Indigo
  • Instrument:
    Includes values for all instruments, except DR/3000 and DR/700
  • Cat. no.:
    CA97026-524
  • Supplier No.:
    2708000

Specifications

About this item

For Ozone Indigo method, MR, 0-.75 mg/L as O₃.

-Fast and Convenient
-Stable

  • Fast and Convenient
  • Stable

Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.

Ordering information: Set of 4 vials.
Customers Who Bought This Also Bought