ContactUsLinkComponent
Testr Paddle Disinfection Pk10
Paddle Tester, Total Aerobic Bacteria/Disinfection Control, Hach
Catalog #: CA97026-326
Supplier:  Hach
CAS Number:  
Testr Paddle Disinfection Pk10
Paddle Tester, Total Aerobic Bacteria/Disinfection Control, Hach
Catalog #: CA97026-326
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Paddle Tester, Total Aerobic Bacteria/Disinfection Control
  • Cat. no.:
    CA97026-326
  • Supplier No.:
    2619510
Specifications
About this item
Hach Paddle Testers allow fast, easy screening of water samples, solid surfaces, and nonviscous liquids where unsanitary conditions may exist.
-Save time and simplify lab and field testing with Hach prepared P/A media
-Report Total Coliform values with EPA accepted Presence-Absence bottles and media
-Simplify routine testing with self-contained, easy-to-use Paddle Testers and BART™ products
  • Save time and simplify lab and field testing with Hach prepared P/A media
  • Report Total Coliform values with EPA accepted PresenceAbsence bottles and media
  • Simplify routine testing with selfcontained, easytouse Paddle Testers and BART™ products
The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a molded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle Tester, Total Aerobic Bacteria/Disinfection Control (Tryptic Soy Agar/DE Neutralizing Agar).
Ordering information: 10/pk.
Customers Who Bought This Also Bought