Order Entry
ContactUsLinkComponent
Buffer Ph4.01 Pwd Plws Pk/15
pH Buffer Powder Pillows, Hach
Catalog #: CA97021-766
Supplier:  Hach
CAS Number:  
Buffer Ph4.01 Pwd Plws Pk/15
pH Buffer Powder Pillows, Hach
Catalog #: CA97021-766
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    4.01
  • Color:
    Red
  • pH Accuracy:
    ±0.02
  • Supplier No.:
    2226995

Specifications

About this item

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination

Fresh and reliable

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

Each powder pillow prepares 50 mL of buffer solutions of respective pH.

Customers Who Bought This Also Bought