Order Entry
ContactUsLinkComponent
RPR BULK KIT - 500-TEST KIT NO 110
Macro-Vue™ RPR Circle Card Test Kits, BD Diagnostics
Catalog #: CA90003-942L
Supplier:  BD
CAS Number:  
RPR BULK KIT - 500-TEST KIT NO 110
Macro-Vue™ RPR Circle Card Test Kits, BD Diagnostics
Catalog #: CA90003-942L
Supplier:  BD
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Macro-Vue™ RPR Circle Card
  • Cat. no.:
    CA90003-942L
  • Supplier No.:
    275005
  • No. of Tests:
    500

Specifications

About this item

These rapid plasma reagin (RPR) test kits are developed for use in large-scale testing in public health and clinical laboratories

Kits use mechanical rotation, antigen suspension, and specially prepared, plastic-coated test cards for the serologic detection of syphilis. The recommended speed for rotation is 100rpm. The rotator should circumscribe a circle approximately 2cm in diameter in the horizontal plane. Refrigeration is recommended for storing the antigen suspension ampules. The disposable BD Dispenstirs™ devices allow the transfer of specimen to the card surface. An antigen dispensing bottle with a needle serves for accurate drop delivery of antigen. Spot cards feature circle spots 18mm in diameter.

Kit contents:
3 mL Antigen
Ampules
10-Spot Cards
Capillaries, 0.05 mL
Antigen Dispensing Bottle Stirrers
20-Gauge Dispensing Needle

Customers Who Bought This Also Bought