Antibiotics and Antimycotics, BD Diagnostics
Catalog #: CA90002-476L
Supplier: BD
CAS Number:
Specifications
- Description:Ciprofloxacin, 5 µg
- Size:—
- Cat. no.:CA90002-476L
- Supplier No.:231658
Specifications
About this item
Ideal for the qualitative susceptibility testing of mycobacteria and related organisms.
- Reliable
- Convenient
- Rapid method
This collection of media products is used for the evaluation of susceptibility of microorganisms to anti-microbial agents.
Packaging: Each test includes 50 susceptibility disks per cartridge.
Customers Who Bought This Also Bought