To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:IntelliCAL PHC301 Standard Refillable pH Electrode, 1 meter cable
- Filling solution required:Yes
- Cat. no.:CA89174-022
- For Use With:HQ11d, HQ30d, HQ40d, HQ411d, HQ430d, HQ440
- pH Range:0 - 14
- Body Material:Zeonor
- Junction:Ceramic
- Supplier No.:PHC30101
Specifications
About this item
IntelliCAL digital probes are factory calibrated and ready to test right out of the box
The IntelliCAL PHC probe is a digital, combination pH probe with a double junction reference and built-in temperature sensor.
IntelliCAL digital probes alert the user when re-calibration is needed, but re-calibration and measurement setting re-entry is not required when moving probes between meters, making these probes ideal for a multi-user environment with multiple HQd series Laboratory and Portable meters. IntelliCAL digital probes additionally provide ultimate traceability in measurement history, storing time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number. No guesswork is needed to provide the most reliable and accurate results with each measurement.