Order Entry
ContactUsLinkComponent
Flask Erlenmeyer Pp 250Ml Cs6
Erlenmeyer Flask, Polypropylene, Dynalon
Catalog #: 30621-006
Supplier:  Dynalon
Flask Erlenmeyer Pp 250Ml Cs6
Erlenmeyer Flask, Polypropylene, Dynalon
Catalog #: 30621-006
Supplier:  Dynalon

Specifications

  • Capacity:
    250 ml
  • Graduation Interval:
    25 ml
  • ST Joint No.:
    19/26
  • Cat. no.:
    30621-006
  • Supplier No.:
    241345

Specifications

About this item

Autoclavable polypropylene Erlenmeyer flask with molded graduations offers excellent chemical resistance.

Graduations are permanently molded into the flask.

Customers Who Bought This Also Bought