Order Entry
ContactUsLinkComponent
Carrier 6-Bottle Econ Hdpe Cs5
Economy Bottle Carrier, Dynalon
Catalog #: 30621-004
Supplier:  Dynalon
Carrier 6-Bottle Econ Hdpe Cs5
Economy Bottle Carrier, Dynalon
Catalog #: 30621-004
Supplier:  Dynalon

Specifications

  • Description:
    HDPE utility carrier for 6 bottles
  • Cat. no.:
    30621-004
  • L×W×H:
    13.56×10.12×11"
  • Weight:
    0.65 lbs.
  • Supplier No.:
    408175

Specifications

About this item

Lightweight, sturdy bottle carrier made from High Density Polyethylene

Transports up to 6 bottles with a diameter of 3 1/2 inches or smaller (usually 250 - 1L sizes). The handle is approximately 6 inches in height.

Customers Who Bought This Also Bought