Fruit Crunchies™ Treats, Certified, Bio-Serv
Catalog #: CA11024-216
Supplier: Bio-Serv
Specifications
- Description:Fruit Crunchies
- Size:Pellets
- Cat. no.:CA11024-216
- Packaging:1 kg/Bag
- Supplier No.:F05798-1
Specifications
About this item
Nutritionally complete 190 mg treat that complements enrichment programs can be used as a reward or foraging treat.
- A medley of apple, grape and orange flavors adds variety to foraging activity
- Excellent source of Vitamin C
- Less labor intensive and more sanitary than perishable fruit
- Sterile version is sterilized by gamma irradiation
- Nutritionally assayed
- Shelf Life: 12 months, cool dry storage
Ingredients: Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Vitamin Mix, Natural and Artificial Flavors, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye.
Caloric value: 0.66 kcal/Pellet
Nutritional ash content: 5.10%
Nutritional carbohydrate content: 52%
Nutritional fat content: 6.30%
Nutritional fibre content: 11.50%
Nutritional moisture content:<10%
Nutritional protein content: 20.20%
Vitamin C: 0.21 mg/Pellet
Certifications: Certified (Contaminant Screened).
Customers Who Bought This Also Bought