Order Entry
ContactUsLinkComponent
ETHIDIUM BRMIDE DESTAI
Destaining Bags, Ethidium Bromide, Thermo Scientific
Catalog #: CAAAJ62931-8U
CAS Number:  
Destaining Bags, Ethidium Bromide, Thermo Scientific
Catalog #: CAAAJ62931-8U
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    25 each
  • Cat. no.:
    CAAAJ62931-8U
  • Supplier No.:
    J62931-8U

Specifications

About this item

Ethidium bromide de-staining bags with activation solution can be used to safely remove ethidium bromide from solution during overnight treatment.

  • Destain overnight with gentle rocking or mixing
  • Soluble in water
  • Stored at ambient temperature

Each bag extracts up to 5 mg of ethidium bromide from solution thus minimizing the exposure of research personnel to this material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration. It provides a safer, more economical means for disposal of hazardous waste.

Customers Who Bought This Also Bought