Potassium phosphate, 0.5 M buffer soln., pH 7.0, Thermo Scientific
Catalog #: CAAAJ60584-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:250 mL
- MDL No.:MFCD00011401
- Cat. no.:CAAAJ60584-AK
- Supplier No.:J60584-AK
Specifications
About this item
Liquid
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Customers Who Bought This Also Bought