Order Entry
ContactUsLinkComponent
Kleenex Hard Roll Towels
Kleenex® Hard Roll Towels, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: 89402-824
Supplier:  Kimberly-Clark
Kleenex Hard Roll Towels
Kleenex® Hard Roll Towels, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: 89402-824
Supplier:  Kimberly-Clark

Specifications

  • Length:
    15.24 m (600")
  • Color:
    White
  • Description:
    Kleenex Hard Roll Towels
  • Cat. no.:
    89402-824
  • Supplier No.:
    11090

Specifications

About this item

Soft feel and quality roll towel for hand drying performance, Kleenex hard roll towels have absorbency pockets absorb lots of water fast, for superior hand drying and less waste.

  • 1.5 core size

Small cases for easy storage and handling.

Certifications: EPA standards, FSC® and EcoLogo® certified.
Customers Who Bought This Also Bought