Element Silicon Carbide Solid
Catalog #: 30620-976
Supplier: DFC Ceramics
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Molecular sieve A4 (0.4 nm, 4 Å) Grade 514, beads Mesh 8 - 12, Supelco®
Supplier: MilliporeSigma
$490.98
Citric acid monohydrate 99.5-100.5% (by acidimetry on anhydrous substance), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
$365.38 to $2,062.18