OIL NOZZLE 0.75 @ 60 DEG CF
Catalog #: CA11023-272
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
$46.00 to $367.00
TURKEY RED BLOOD CELLS 10% Washed Pooled Cells, Rockland Immunochemicals
Supplier: Rockland Immunochemical
$597.24
Chloroform-[D₁] (99.8% D) (Isotopic) +1% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
$124.00
Chlorate (ClO₃)- Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR International
$194.70