To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:F4/80
- Clonality:Monoclonal
- Clone:BM8
- Gene ID:13733
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:F4/80
- Conjugation:Brilliant Violet® 421
- ImmunoChemistry:Yes
- Size:500 µL
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Antigen synonyms:Ly71|EMR1|ADGRE1
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:Murine macrophages
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 421™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 421™ and unconjugated antibody.
- Cat. no.:CA123132-BL
- Supplier No.:123132
Specifications
About this item
Brilliant Violet 421™ anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 50 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For immunohistochemical staining on frozen tissue sections, the suggested use of this reagent is 2.0 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse