Order Entry
ContactUsLinkComponent
HALOACETIC ACIDS (HAA)
Water Supply Standard, Haloacetic Acids, ERA
Catalog #: CA11022-698
Supplier:  ERA
CAS Number:  
Water Supply Standard, Haloacetic Acids, ERA
Catalog #: CA11022-698
Supplier:  ERA
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Certified Reference Material
  • Concentration:
    5-50 µg/L
  • Cat. no.:
    CA11022-698
  • Supplier No.:
    684
  • Parameters:
    Haloacetic Acids (HAA)

Specifications

About this item

One 2 mL flame-sealed ampule yields up to 2 L of this water supply, haloacetic acids (HAA) standard after dilution

Bromochloroacetic Acid, Dibromoacetic Acid, Dichloroacetic Acid, Monobromoacetic Acid, Monochloroacetic Acid, Trichloroacetic Acid

Use with EPA method 552. This standard includes all of the analytes listed below at 5 to 50 µg/L.

Customers Who Bought This Also Bought