Glass Disruptor Beads, Chemglass
Catalog #: 30623-118
Supplier: Chemglass
Small Business Enterprise
Specifications
- Ø:0.1 mm
- Description:Glass disruptor beads for bacteria
- Size:8 oz.
- Supplier No.:CLS-1835-BG1
Specifications
About this item
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$233.35 to $2,174.33
Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Supplier: BDH
$720.66 to $3,438.57