ChemRxnHub™ Systems, Jacketed, 2000 ml, Jacketed Lab Reactor, Chemglass
Catalog #: 30622-574
Supplier: Chemglass
Small Business Enterprise
Specifications
- Description:ChemRxnHub Process System Complete with Glassware, Stand, and OHS 100 Advance Motor
- Cat. no.:30622-574
- Supplier No.:CG-1949-X-2L
Specifications
About this item
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.
Customers Who Bought This Also Bought