Alpine Fire Assay Cupels, MgO, Anachemia
Catalog #: CA11022-138
Supplier: Anachemia Mining
Specifications
- I.D.:30.0 mm
- O.D.:40.0 mm
- Description:6A Tall
- Height:32.0 mm
- Cat. no.:CA11022-138
- Top Diameter:40 mm
- Absorbing lead:64 g
- Bottom Diameter:30 mm
- Depth:11.0 mm
- Supplier No.:029-730-07
Specifications
About this item
Alpine cupels are made from high purity magnesium oxide and offer superior mechanical strength free from cracking and pitting
- High-purity material
- Superior mechanical strength
- Consistent low losses
These cupels offer consistent low losses of precious metals and are continually tested to ensure product quality remains at the highest level.
Customers Who Bought This Also Bought