Water Pollution Standard, Minerals, ERA (Waters)
Catalog # CA11020-088
Supplier: ERA
CAS Number:
Specifications
- Description:Certified Reference Material
- Size:500 ml
- Sodium:10-100 mg/L
- Potassium:4-40 mg/L
- Fluoride:0.4-4 mg/L
- Specific conductance:200-1200 μmhos/cm at 25 °C
- Alkalinity:25-400 mg/L (as CaCO3)
- Chloride:35-275 mg/L
- Sulphate:5-125 mg/L
- TDS:140-800 mg/L at 180 °C
- Total solids:140-800 mg/L at 105 °C
- Cat. no.:CA11020-088
- Supplier No.:506
- Parameters:Minerals
Specifications
About this item
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 500 mL whole-volume bottle of this water pollution, minerals standard is ready to analyze.
Specific conductance: 200 - 1200 µmhos/cm (at 25 °C)
Frequently Bought Together