TILE HEARTH MODIFIED FH-207 63
Catalog #: 30617-916
Supplier: DFC Ceramics
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Rubidium hydroxide 50% (w/w) in aqueous solution , 99.6% (metals basis)
Supplier: Thermo Scientific Chemicals
$212.00
Conductivity Standards, Potassium Chloride (KCl) Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
$58.00