BOARD TRAY 19.4X12X36IN
Catalog #: 30617-914
Supplier: DFC Ceramics
Customers Who Bought This Also Bought
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents, Thermo Scientific
Supplier: Thermo Fisher Scientific
$180.89
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Ammonium carbonate 30,0% (NH₃ basis), lumps, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$267.72 to $843.18