T/C PROT TUBE
Catalog #: 30617-878
Supplier: DFC Ceramics
Customers Who Bought This Also Bought
Sodium hydroxide 10.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$82.35 to $926.81
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$619.49 to $3,535.84
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$330.55 to $1,952.15