ContactUsLinkComponent
CUVETTE CAPS SQ LDPE CS1000
Cuvette Cap, Square, Dynalon
Catalog #: 30620-274
Supplier:  Dynalon
CUVETTE CAPS SQ LDPE CS1000
Cuvette Cap, Square, Dynalon
Catalog #: 30620-274
Supplier:  Dynalon
Specifications
  • Length:
    1/2"
  • Width:
    1/2"
  • Height:
    1/4"
  • Cat. no.:
    30620-274
  • Weight:
    0.00062 lbs
  • Supplier No.:
    402364
Specifications
About this item
Cuvette caps, polyethylene dust covers fit most standard cuvettes.
Flexible fins on cap allow for tight fit that will not damage the cuvette. Caps are 19 mm in depth.
Customers Who Bought This Also Bought