SP Bel-Art Poxygrid® Test Tube Racks, Bel-Art Products, a part of SP
Catalog #: 30617-642
Supplier: Bel-Art Products, a Part of SP
Specifications
- Dimensions:29.3L×12.8W×8.3H cm (119/16×51/16×31/4")
- Color:Blue
- No. of Holes:40 (4×10)
- Cat. no.:30617-642
- For Tube O.D.:20 - 25 mm
- Supplier No.:187720001
Specifications
About this item
Chemical- and corrosion-resistant steel wire racks are coated with heat-cured epoxy.
- Epoxy coatings are unaffected by organic solvents, salts, weak acids, and weak alkalis
- Slotted grid bottom and reinforced edges
- Autoclavable
They will withstand a temperature range of -223 to 121 °C (-369 to 250 °F) and can be autoclaved.
All racks have square holes, three decks, a slotted grid bottom, and reinforced edges.
Orange racks can be used for biohazard warning, yellow for radioactive warning.
Customers Who Bought This Also Bought