VIAL KIT POLYSTYREN 10ML PK100
Catalog #: CA11019-468
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90