Lead, foil
Catalog #: CA11019-412
Supplier: ALPHA ASSEMBLY SOLUTIONS CA
CAS Number: 7439-92-1
Specifications
- Packaging:Roll
- Size:5 lbs.
- Cat. no.:CA11019-412
- Supplier No.:LEADFOIL
Specifications
Specification Test Results
- Thickness:0.1 mm
Specification Test Results
About this item
Boiling Pt:1740 °C (1013 hPa)
Melting Pt:327 °C
Density:11.3437 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 1740 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 327 °C
:Flash Pt:
:Flash Pt:
:M.W. 207.2 g/mol
Merck Index:12,05410
:Lead
EINECS:231-100-4
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Customers Who Bought This Also Bought